1,2-Diiodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001039-5G 001039-25G
FLUO001039-5GEA 0 GBP
FLUO001039-5G FLUO001039-25G
1,2-Diiodobenzene
1,2-Diiodobenzene
Formula: C₆H₄I₂
Boiling Pt: 285 °C (1013 hPa)
Density: 2,76 g/cm³ (0 °C)
MDL Number: MFCD00001038
CAS Number: 615-42-9
EINECS: 210-425-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR