4-Fluorothiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol

001625-25G 001625-100G 001625-250G
FLUO001625-25GEA 0 GBP
FLUO001625-25G FLUO001625-100G FLUO001625-250G
4-Fluorothiophenol
4-Fluorothiophenol
Formula: C₆H₅FS
Boiling Pt: 164 °C (1013 hPa)
Density: 1.1996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004846
CAS Number: 371-42-6
EINECS: 206-737-6
UN: 3071
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR