2,3,6-Trifluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001850-5G 001850-25G
FLUO001850-5GEA 0 GBP
FLUO001850-5G FLUO001850-25G
2,3,6-Trifluorophenol
2,3,6-Trifluorophenol
Formula: C₆H₃F₃O
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: 33…37 °C
MDL Number: MFCD00061214
CAS Number: 113798-74-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR