2,3,5,6-Tetrafluorophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001820-100G
001820-25G
FLUO001820-100GEA
0
GBP
FLUO001820-100G
FLUO001820-25G
2,3,5,6-Tetrafluorophenol
2,3,5,6-Tetrafluorophenol
Formula:
C₆H₂F₄O Boiling Pt: 140 °C (1013 hPa) Melting Pt: 30 °C |
MDL Number:
MFCD00002157 CAS Number: 769-39-1 EINECS: 212-209-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...