3-Fluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002173-25G 002173-100G 002173-500G
FLUO002173-25GEA 0 GBP
FLUO002173-25G FLUO002173-100G FLUO002173-500G
3-Fluorophenol
3-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: 13.7 °C
Density: 1.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002254
CAS Number: 372-20-3
EINECS: 206-748-6
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR