5-Fluoro-2-nitrophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001470-25G
001470-100G
001470-250G
FLUO001470-25GEA
0
GBP
FLUO001470-25G
FLUO001470-100G
FLUO001470-250G
5-Fluoro-2-nitrophenol
5-Fluoro-2-nitrophenol
Formula:
C₆H₄FNO₃ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 36…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007107 CAS Number: 446-36-6 EINECS: 207-168-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...