2,4-Difluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001926-25G 001926-100G 001926-250G
FLUO001926-25GEA 0 GBP
FLUO001926-25G FLUO001926-100G FLUO001926-250G
2,4-Difluorophenol
2,4-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00009715
CAS Number: 367-27-1
EINECS: 206-688-0
UN: 2921
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR