2,6-Difluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001418-5G 001418-25G 001418-100G
FLUO001418-5GEA 0 GBP
FLUO001418-5G FLUO001418-25G FLUO001418-100G
2,6-Difluorophenol
2,6-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 66 °C (20 mmHg)
Melting Pt: 46 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002158
CAS Number: 28177-48-2
EINECS: 248-884-9
UN: 2921
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR