4-(Trifluoromethoxy)anisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007532-1G 007532-5G 007532-25G
FLUO007532-1GEA 0 GBP
FLUO007532-1G FLUO007532-5G FLUO007532-25G
4-(Trifluoromethoxy)anisole
4-(Trifluoromethoxy)anisole
Formula: C₈H₇F₃O₂
Boiling Pt: 163…165 °C (1013 hPa)
Density: 1.272 g/cm³ (20 °C)
MDL Number: MFCD00216942
CAS Number: 710-18-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR