Pentafluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006946-5G 006946-25G 006946-100G
FLUO006946-5GEA 0 GBP
FLUO006946-5G FLUO006946-25G FLUO006946-100G
Pentafluoroanisole
Pentafluoroanisole
Formula: C₇H₃F₅O
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –37 °C
Density: 1.496 g/cm³ (20 °C)
MDL Number: MFCD00000289
CAS Number: 389-40-2
EINECS: 206-866-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR