2,3,6-Trifluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005850-5G 005850-1G
FLUO005850-5GEA 0 GBP
FLUO005850-5G FLUO005850-1G
2,3,6-Trifluoroanisole
2,3,6-Trifluoroanisole
Formula: C₇H₅F₃O
Boiling Pt: 146 °C (1013 hPa)
MDL Number: MFCD01631501
CAS Number: 4920-34-7
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR