2,3,5,6-Tetrafluoroanisole ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B24275.06
B24275.14
B24275.06EA
47.3
GBP
B24275.06
B24275.14
2,3,5,6-Tetrafluoroanisole ≥97%
2,3,5,6-Tetrafluoroanisole
Purity: 97%
CAS No.: 2324-98-3
Molecular Formula: C7H4F4O
Formula Weight: 180.10
Flash Point: 41°(107°F)
Refractive Index: 1.4280
Boiling Point: 144-145°
UN No.: 1993
MDL No.: MFCD00000311
Formula:
C₇H₄F₄O MW: 180.1 g/mol Boiling Pt: 144…145 °C Density: 1.293 Flash Pt: 41 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000311 CAS Number: 2324-98-3 EINECS: 219-037-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...