2-Fluoroanisole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002694-100G
002694-500G
FLUO002694-100GEA
0
GBP
FLUO002694-100G
FLUO002694-500G
2-Fluoroanisole
2-Fluoroanisole
Formula:
C₇H₇FO Boiling Pt: 154 °C (1013 hPa) Melting Pt: –39 °C Density: 1.124 g/cm³ (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000316 CAS Number: 321-28-8 EINECS: 206-284-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...