2,5-Difluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,4-Difluoro-2-methoxybenzene

002347-100G 002347-25G
FLUO002347-100GEA 0 GBP
FLUO002347-100G FLUO002347-25G
2,5-Difluoroanisole
2,5-Difluoroanisole
Formula: C₇H₆F₂O
Boiling Pt: 72…73 °C (25 mmHg)
MDL Number: MFCD00042561
CAS Number: 75626-17-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR