2,4-Difluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,4-Difluoro-1-methoxybenzene

002346-25G 002346-100G
FLUO002346-25GEA 0 GBP
FLUO002346-25G FLUO002346-100G
2,4-Difluoroanisole
2,4-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 53 °C (1013 hPa)
Density: 1.2513 g/cm³ (20.7 °C)
MDL Number: MFCD00010731
CAS Number: 452-10-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR