3,4-Difluoroanisole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
004776-25G
004776-10G
FLUO004776-25GEA
0
GBP
FLUO004776-25G
FLUO004776-10G
3,4-Difluoroanisole
3,4-Difluoroanisole
Formula:
C₇H₆F₂O Boiling Pt: 139 °C (1013 hPa) Melting Pt: –30 °C Density: 1.23 g/cm³ (20 °C) |
MDL Number:
MFCD00010730 CAS Number: 115144-40-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...