Sanger's reagent

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene

002693-25G 002693-100G
FLUO002693-25GEA 0 GBP
FLUO002693-25G FLUO002693-100G
Sanger's reagent
Sanger's reagent
Formula: C₆H₃FN₂O₄
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 24…26 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007056
CAS Number: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,04199

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR