1,2,4-Trifluoro-5-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4,5-Trifluoronitrobenzene

002363-25G 002363-100G
FLUO002363-25GEA 0 GBP
FLUO002363-25G FLUO002363-100G
1,2,4-Trifluoro-5-nitrobenzene
1,2,4-Trifluoro-5-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: –11 °C
Density: 1.54 g/cm³ (20 °C)
MDL Number: MFCD00007051
CAS Number: 2105-61-5
EINECS: 218-281-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR