1-Fluoro-4-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002698-250G 002698-1KG 002698-5KG
FLUO002698-250GEA 0 GBP
FLUO002698-250G FLUO002698-1KG FLUO002698-5KG
1-Fluoro-4-nitrobenzene
1-Fluoro-4-nitrobenzene
Formula: C₆H₄FNO₂
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.326 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007282
CAS Number: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR