1-Fluoro-3-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Fluoronitrobenzene
001678-25G
001678-100G
001678-250G
FLUO001678-25GEA
0
GBP
FLUO001678-25G
FLUO001678-100G
FLUO001678-250G
1-Fluoro-3-nitrobenzene
1-Fluoro-3-nitrobenzene
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Boiling Pt: 205 Melting Pt: 1.7 Density: 1.33 g/cm³ (20 °C) Flash Pt: 76 Storage Temperature: Ambient |
MDL Number:
MFCD00007196 CAS Number: 402-67-5 EINECS: 206-953-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...