1-Fluoro-2-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
o-Fluoronitrobenzene, 2-Fluoronitrobenzene
001674-250G
001674-1KG
001674-5KG
FLUO001674-250GEA
0
GBP
FLUO001674-250G
FLUO001674-1KG
FLUO001674-5KG
1-Fluoro-2-nitrobenzene
1-Fluoro-2-nitrobenzene
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Boiling Pt: 212 Melting Pt: –8 °C Density: 1.33 g/cm³ (20 °C) Flash Pt: 94 Storage Temperature: Ambient |
MDL Number:
MFCD00007048 CAS Number: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...