1,4-Difluoro-2-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,5-Difluoronitrobenzene
001942-100G
001942-500G
001942-50G
FLUO001942-100GEA
0
GBP
FLUO001942-100G
FLUO001942-500G
FLUO001942-50G
1,4-Difluoro-2-nitrobenzene
1,4-Difluoro-2-nitrobenzene
Formula:
C₆H₃F₂NO₂ Boiling Pt: 207 °C (1013 hPa) Melting Pt: –11.7 °C Density: 1.4671 g/cm³ (17 °C) |
MDL Number:
MFCD00007054 CAS Number: 364-74-9 EINECS: 206-663-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...