1,3-Difluoro-5-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
019498-5G
019498-100G
019498-25G
FLUO019498-5GEA
0
GBP
FLUO019498-5G
FLUO019498-100G
FLUO019498-25G
1,3-Difluoro-5-nitrobenzene
1,3-Difluoro-5-nitrobenzene
Formula:
C₆H₃F₂NO₂ MW: 159.09 g/mol |
MDL Number:
MFCD00012142 CAS Number: 2265-94-3 EINECS: 218-867-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...