1,3-Difluoro-4-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene
001432-250G
001432-1KG
FLUO001432-250GEA
0
GBP
FLUO001432-250G
FLUO001432-1KG
1,3-Difluoro-4-nitrobenzene
1,3-Difluoro-4-nitrobenzene
Formula:
C₆H₃F₂NO₂ Boiling Pt: 203 °C (1013 hPa) Melting Pt: 10 °C Density: 1.4571 g/cm³ (14 °C) |
MDL Number:
MFCD00007050 CAS Number: 446-35-5 EINECS: 207-167-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...