4-Fluoro-3-nitroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001697-25G
001697-100G
001697-500G
FLUO001697-25GEA
0
GBP
FLUO001697-25G
FLUO001697-100G
FLUO001697-500G
4-Fluoro-3-nitroaniline
4-Fluoro-3-nitroaniline
Formula:
C₆H₅FN₂O₂ Melting Pt: 96…98 °C Density: 0,43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007833 CAS Number: 364-76-1 EINECS: 206-665-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...