4-Fluoro-2-nitroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001688-25G
001688-100G
FLUO001688-25GEA
0
GBP
FLUO001688-25G
FLUO001688-100G
4-Fluoro-2-nitroaniline
4-Fluoro-2-nitroaniline
Formula:
C₆H₅FN₂O₂ Melting Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007830 CAS Number: 364-78-3 EINECS: 206-666-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...