2-Fluoro-5-nitroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003230-25G 003230-100G
FLUO003230-25GEA 0 GBP
FLUO003230-25G FLUO003230-100G
2-Fluoro-5-nitroaniline
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR