2,3,4,5,6-Pentafluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Aminopentafluorobenzene, Pentafluoroaniline

002834-25G 002834-100G
FLUO002834-25GEA 0 GBP
FLUO002834-25G FLUO002834-100G
2,3,4,5,6-Pentafluoroaniline
2,3,4,5,6-Pentafluoroaniline
Formula: C₆H₂F₅N
MW: 183.08 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: 36…37 °C
Density: 1,744 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007643
CAS Number: 771-60-8
EINECS: 212-234-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR