4-Fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001439-100G 001439-500G 001439-2KG
FLUO001439-100GEA 0 GBP
FLUO001439-100G FLUO001439-500G FLUO001439-2KG
4-Fluoroaniline
4-Fluoroaniline
Formula: C₆H₆FN
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR