3-Fluoroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001438-100G
001438-1KG
001438-500G
FLUO001438-100GEA
0
GBP
FLUO001438-100G
FLUO001438-1KG
FLUO001438-500G
3-Fluoroaniline
3-Fluoroaniline
Formula:
C₆H₆FN Boiling Pt: 186 °C (1013 hPa) Melting Pt: –2 °C Density: 1.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007758 CAS Number: 372-19-0 EINECS: 206-747-0 UN: 2941 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...