3,5-Difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001690-5G 001690-25G 001690-100G
FLUO001690-5GEA 0 GBP
FLUO001690-5G FLUO001690-25G FLUO001690-100G
3,5-Difluoroaniline
3,5-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 37…41 °C
Density: 1.295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR