3,4-Difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001459-100G 001459-500G 001459-25G
FLUO001459-100GEA 0 GBP
FLUO001459-100G FLUO001459-500G FLUO001459-25G
3,4-Difluoroaniline
3,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 205
Melting Pt: 22
Density: 1.302 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007761
CAS Number: 3863-11-4
EINECS: 223-381-7
UN: 2941
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR