2,6-Difluoroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001458-100G
001458-500G
001458-25G
FLUO001458-100GEA
0
GBP
FLUO001458-100G
FLUO001458-500G
FLUO001458-25G
2,6-Difluoroaniline
2,6-Difluoroaniline
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 51 °C (1013 hPa) Density: 1.199 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007655 CAS Number: 5509-65-9 EINECS: 226-853-0 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...