2,5-Difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001931-500G 001931-100G 001931-25G
FLUO001931-500GEA 0 GBP
FLUO001931-500G FLUO001931-100G FLUO001931-25G
2,5-Difluoroaniline
2,5-Difluoroaniline
Formula: C₆H₅F₂N
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.285 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007651
CAS Number: 367-30-6
EINECS: 206-690-1
UN: 2941
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR