2,4-Difluoroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001428-100G
001428-500G
001428-2.5KG
FLUO001428-100GEA
0
GBP
FLUO001428-100G
FLUO001428-500G
FLUO001428-2.5KG
2,4-Difluoroaniline
2,4-Difluoroaniline
Formula:
C₆H₅F₂N Boiling Pt: 170 °C (1013 hPa) Melting Pt: –7 °C Density: 1.268 g/cm³ (20 °C) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007648 CAS Number: 367-25-9 EINECS: 206-687-5 UN: 2941 ADR: 6.1,III Merck Index: 13,03171 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...