2,3,5,6-Tetrafluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001355-5G 001355-25G 001355-10G
FLUO001355-5GEA 0 GBP
FLUO001355-5G FLUO001355-25G FLUO001355-10G
2,3,5,6-Tetrafluoroaniline
2,3,5,6-Tetrafluoroaniline
Formula: C₆H₃F₄N
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 31…32 °C
Density: 1.744 g/cm³ (20 °C)
MDL Number: MFCD00007644
CAS Number: 700-17-4
EINECS: 211-840-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR