4-(Trifluoromethoxy)benzoic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

FLUO001545-100GEA 0 GBP
FLUO001545-100G FLUO001545-10G FLUO001545-25G
4-(Trifluoromethoxy)benzoic acid
4-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 153.4 °C
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR