4-(Difluoromethoxy)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

004831-5G 004831-10G 004831-1G
FLUO004831-5GEA 0 GBP
FLUO004831-5G FLUO004831-10G FLUO004831-1G
4-(Difluoromethoxy)benzoic acid
4-(Difluoromethoxy)benzoic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
MDL Number: MFCD00221454
CAS Number: 4837-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR