3-(Trifluoromethoxy)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001544-1G 001544-5G 001544-25G
FLUO001544-1GEA 0 GBP
FLUO001544-1G FLUO001544-5G FLUO001544-25G
3-(Trifluoromethoxy)benzoic acid
3-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 91…92 °C
MDL Number: MFCD00041501
CAS Number: 1014-81-9
EINECS: 213-802-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR