6-Fluoroanthranilic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003980-5G
003980-25G
FLUO003980-5GEA
0
GBP
FLUO003980-5G
FLUO003980-25G
6-Fluoroanthranilic acid
6-Fluoroanthranilic acid
Formula:
C₇H₆FNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 165…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067781 CAS Number: 434-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...