5-Fluoroanthranilic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003126-25G 003126-5G 003126-100G
FLUO003126-25GEA 0 GBP
FLUO003126-25G FLUO003126-5G FLUO003126-100G
5-Fluoroanthranilic acid
5-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00055566
CAS Number: 446-08-2
EINECS: 207-159-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR