3-Fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002167-25G 002167-100G 002167-500G
FLUO002167-25GEA 0 GBP
FLUO002167-25G FLUO002167-100G FLUO002167-500G
3-Fluorobenzoic acid
3-Fluorobenzoic acid
Formula: C₇H₅FO₂
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR