3,5-Difluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001417-25G 001417-100G 001417-500G
FLUO001417-25GEA 0 GBP
FLUO001417-25G FLUO001417-100G FLUO001417-500G
3,5-Difluorobenzoic acid
3,5-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00010323
CAS Number: 455-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR