3,4-Difluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002080-25G 002080-100G 002080-500G
FLUO002080-25GEA 0 GBP
FLUO002080-25G FLUO002080-100G FLUO002080-500G
3,4-Difluorobenzoic acid
3,4-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 119…122 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011672
CAS Number: 455-86-7
EINECS: 207-249-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR