3,4,5-Trifluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002067-5G
002067-25G
FLUO002067-5GEA
0
GBP
FLUO002067-5G
FLUO002067-25G
3,4,5-Trifluorobenzoic acid
3,4,5-Trifluorobenzoic acid
Formula:
C₇H₃F₃O₂ MW: 176.09 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 97…99 °C |
MDL Number:
MFCD00074962 CAS Number: 121602-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...