2,5-Difluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001934-25G
001934-100G
001934-250G
FLUO001934-25GEA
0
GBP
FLUO001934-25G
FLUO001934-100G
FLUO001934-250G
2,5-Difluorobenzoic acid
2,5-Difluorobenzoic acid
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol Boiling Pt: 242 Melting Pt: 131…132 °C |
MDL Number:
MFCD00002410 CAS Number: 2991-28-8 EINECS: 221-060-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...