2,4-Difluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001917-25G 001917-100G 001917-500G
FLUO001917-25GEA 0 GBP
FLUO001917-25G FLUO001917-100G FLUO001917-500G
2,4-Difluorobenzoic acid
2,4-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00011670
CAS Number: 1583-58-0
EINECS: 216-430-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR