2,4,5-Trifluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001635-25G
001635-100G
001635-250G
FLUO001635-25GEA
0
GBP
FLUO001635-25G
FLUO001635-100G
FLUO001635-250G
2,4,5-Trifluorobenzoic acid
2,4,5-Trifluorobenzoic acid
Formula:
C₇H₃F₃O₂ Boiling Pt: 237 °C (1013 hPa) Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013306 CAS Number: 446-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...