4-Fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: p-Fluorobenzonitrile

002237-100G 002237-500G
FLUO002237-100GEA 0 GBP
FLUO002237-100G FLUO002237-500G
4-Fluorobenzonitrile
4-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: 37…40 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001812
CAS Number: 1194-02-1
EINECS: 214-784-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR