2-Fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001656-100G 001656-500G 001656-1KG
FLUO001656-100GEA 0 GBP
FLUO001656-100G FLUO001656-500G FLUO001656-1KG
2-Fluorobenzonitrile
2-Fluorobenzonitrile
Formula: C₇H₄FN
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 35…37 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001773
CAS Number: 394-47-8
EINECS: 206-897-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR